Merck
  • Stepwise remodeling and subcompartment formation in individual vesicles by three ESCRT-III proteins.
Stepwise remodeling and subcompartment formation in individual vesicles by three ESCRT-III proteins.
iScience (2023-01-03)
Yunuen Avalos-Padilla, Vasil N Georgiev, Eleanor Ewins, Tom Robinson, Esther Orozco, Reinhard Lipowsky, Rumiana Dimova
ABSTRACT
The endosomal sorting complex required for transport (ESCRT) is a multi-protein machinery involved in several membrane remodeling processes. Different approaches have been used to resolve how ESCRT proteins scission membranes. However, the underlying mechanisms generating membrane deformations are still a matter of debate. Here, giant unilamellar vesicles, microfluidic technology, and micropipette aspiration are combined to continuously follow the ESCRT-III-mediated membrane remodeling on the single-vesicle level for the first time. With this approach, we identify different mechanisms by which a minimal set of three ESCRT-III proteins from Entamoeba histolytica reshape the membrane. These proteins modulate the membrane stiffness and spontaneous curvature to regulate bud size and generate intraluminal vesicles even in the absence of ATP. We demonstrate that the bud stability depends on the protein concentration and membrane tension. The approaches introduced here should open the road to diverse applications in synthetic biology for establishing artificial cells with several membrane compartments.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
5R-PLEX beads, for PCR purification
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane