Merck
  • Brain Testosterone-CYP1B1 (Cytochrome P450 1B1) Generated Metabolite 6β-Hydroxytestosterone Promotes Neurogenic Hypertension and Inflammation.
Brain Testosterone-CYP1B1 (Cytochrome P450 1B1) Generated Metabolite 6β-Hydroxytestosterone Promotes Neurogenic Hypertension and Inflammation.
Hypertension (Dallas, Tex. : 1979) (2020-08-07)
Purnima Singh, Shubha Ranjan Dutta, Chi Young Song, SaeRam Oh, Frank J Gonzalez, Kafait U Malik
ABSTRACT
Previously, we showed that peripheral administration of 6β-hydroxytestosterone, a CYP1B1 (cytochrome P450 1B1)-generated metabolite of testosterone, promotes angiotensin II-induced hypertension in male mice. However, the site of action and the underlying mechanism by which 6β-hydroxytestosterone contributes to angiotensin II-induced hypertension is not known. Angiotensin II increases blood pressure by its central action, and CYP1B1 is expressed in the brain. This study was conducted to determine whether testosterone-CYP1B1 generated metabolite 6β-hydroxytestosterone locally in the brain promotes the effect of systemic angiotensin II to produce hypertension in male mice. Central CYP1B1 knockdown in wild-type (Cyp1b1+/+) mice by intracerebroventricular-adenovirus-GFP (green fluorescence protein)-CYP1B1-short hairpin (sh)RNA attenuated, whereas reconstitution of CYP1B1 by adenovirus-GFP-CYP1B1-DNA in the paraventricular nucleus but not in subfornical organ in Cyp1b1-/- mice restored angiotensin II-induced increase in systolic blood pressure measured by tail-cuff. Intracerebroventricular-testosterone in orchidectomized (Orchi)-Cyp1b1+/+ but not in Orchi-Cyp1b1-/-, and intracerebroventricular-6β-hydroxytestosterone in the Orchi-Cyp1b1-/- mice restored the angiotensin II-induced: (1) increase in mean arterial pressure measured by radiotelemetry, and autonomic imbalance; (2) reactive oxygen species production in the subfornical organ and paraventricular nucleus; (3) activation of microglia and astrocyte, and neuroinflammation in the paraventricular nucleus. The effect of intracerebroventricular-6β-hydroxytestosterone to restore the angiotensin II-induced increase in mean arterial pressure and autonomic imbalance in Orchi-Cyp1b1-/- mice was inhibited by intracerebroventricular-small interfering (si)RNA-androgen receptor (AR) and GPRC6A (G protein-coupled receptor C6A). These data suggest that testosterone-CYP1B1-generated metabolite 6β-hydroxytestosterone, most likely in the paraventricular nucleus via AR and GPRC6A, contributes to angiotensin II-induced hypertension and neuroinflammation in male mice.
Sigma-Aldrich
6β-Hydroxytestosterone, ≥97% (HPLC)