Merck
  • Functional and structural changes resulting from strain differences in the rat model of oxygen-induced retinopathy.
Functional and structural changes resulting from strain differences in the rat model of oxygen-induced retinopathy.
Investigative ophthalmology & visual science (2009-01-27)
Allison Lindsay Dorfman, Anna Polosa, Sandrine Joly, Sylvain Chemtob, Pierre Lachapelle
ABSTRACT
Results of studies that compared the racial incidence of retinopathy of prematurity (ROP) suggested that ocular pigmentation might offer protection against the development of severe ROP. The structural and functional consequences of postnatal hyperoxia (oxygen-induced retinopathy; OIR) were compared in albino Sprague-Dawley (SD) and pigmented Long-Evans (LE) rats to verify this finding. Newborn rats were exposed to 80% O(2) during selected postnatal day intervals. The severity of the OIR was determined by examining retinal flatmounts (retinal vasculature assessment), protein level quantification and cellular localization of fibroblast growth factor (FGF)-2 and ciliary neurotrophic factor (CNTF; Western blot analysis and immunohistochemistry, respectively), retinal histology, and photopic and scotopic electroretinograms (ERGs). Irrespective of the parameter considered, structural and functional deficits resulting from postnatal hyperoxia were significantly more pronounced in LE rats. Although FGF-2 levels in LE rats had a tendency to increase after hyperoxia compared with normoxic littermates, it did not reach statistical significance. A similar finding was observed in SD rats. Of interest, however, baseline levels of FGF-2 were approximately four to five times higher in SD rats than in LE rats. There was a similar, hyperoxia-induced increase in CNTF levels between SD and LE rats. The findings suggest an increased susceptibility of newborn LE rats to postnatal hyperoxia in comparison with SD rats. Whether a pro-oxidant rather than antioxidant role of melanin or other genetic factors can explain these differences in oxygen susceptibility of the animal model of this retinopathy, remains to be determined.
Sigma-Aldrich
Anti-Ciliary Neurotrophic Factor Antibody, clone 4-68, clone 4-68, Chemicon®, from mouse
Sigma-Aldrich
Anti-FGF-2/basic FGF Antibody, clone bFM-2, clone bFM-2, Upstate®, from mouse