Merck
  • Quantitative spectroscopic imaging for non-invasive early cancer detection.
Quantitative spectroscopic imaging for non-invasive early cancer detection.
Optics express (2008-10-01)
Chung-Chieh Yu, Condon Lau, Geoffrey O'Donoghue, Jelena Mirkovic, Sasha McGee, Luis Galindo, Alphi Elackattu, Elizabeth Stier, Gregory Grillone, Kamran Badizadegan, Ramachandra R Dasari, Michael S Feld
ABSTRACT
We report a fully quantitative spectroscopy imaging instrument for wide area detection of early cancer (dysplasia). This instrument provides quantitative maps of tissue biochemistry and morphology, making it a potentially powerful surveillance tool for objective early cancer detection. We describe the design, construction, calibration, and first clinical application of this new system. We demonstrate its accuracy using physical tissue models. We validate its diagnostic ability on a resected colon adenoma, and demonstrate feasibility of in vivo imaging in the oral cavity.
Sigma-Aldrich
Hemoglobin human, lyophilized powder
Sigma-Aldrich
Hemoglobin equine, lyophilized powder
Sigma-Aldrich
Hemoglobin porcine, lyophilized powder