Merck
  • Design, synthesis and biological evaluation of naphthalimide-based fluorescent probes for α1-adrenergic receptors.
Design, synthesis and biological evaluation of naphthalimide-based fluorescent probes for α1-adrenergic receptors.
Drug discoveries & therapeutics (2014-03-22)
Wei Zhang, Laizhong Chen, Zhao Ma, Lupei Du, Minyong Li
ABSTRACT
α1-Adrenergic receptors (α1-ARs), as one of the most important members of G protein-coupled receptors (GPCRs), can mediate lots of physiological responses of the sympathetic nervous system. Until now, α1-ARs have been divided into at least three subtypes, α1A, α1B, and α1D, which distribute in various tissues and organs. In this research, we designed and synthesized several napthalimide-based small-molecule fluorescent probes for α1-ARs, which mainly contained two parts: the pharmacophore (quinazoline and phenylpiperazine) that binds to α1-ARs and the fluorophore (naphthalimide) that labels the receptors with fluorescent properties. Moreover, some of these compounds demonstrated potent affinity to α1-ARs and cell imaging potential.
Sigma-Aldrich
Phentolamine hydrochloride, ≥98% (TLC), powder