Merck
  • New phosphosite-specific antibodies to unravel the role of GRK phosphorylation in dopamine D2 receptor regulation and signaling.
New phosphosite-specific antibodies to unravel the role of GRK phosphorylation in dopamine D2 receptor regulation and signaling.
Scientific reports (2021-04-17)
Anika Mann, Alastair C Keen, Hanka Mark, Pooja Dasgupta, Jonathan A Javitch, Meritxell Canals, Stefan Schulz, J Robert Lane
ABSTRACT
The dopamine D2 receptor (D2R) is the target of drugs used to treat the symptoms of Parkinson's disease and schizophrenia. The D2R is regulated through its interaction with and phosphorylation by G protein receptor kinases (GRKs) and interaction with arrestins. More recently, D2R arrestin-mediated signaling has been shown to have distinct physiological functions to those of G protein signalling. Relatively little is known regarding the patterns of D2R phosphorylation that might control these processes. We aimed to generate antibodies specific for intracellular D2R phosphorylation sites to facilitate the investigation of these mechanisms. We synthesised double phosphorylated peptides corresponding to regions within intracellular loop 3 of the hD2R and used them to raise phosphosite-specific antibodies to capture a broad screen of GRK-mediated phosphorylation. We identify an antibody specific to a GRK2/3 phosphorylation site in intracellular loop 3 of the D2R. We compared measurements of D2R phosphorylation with other measurements of D2R signalling to profile selected D2R agonists including previously described biased agonists. These studies demonstrate the utility of novel phosphosite-specific antibodies to investigate D2R regulation and signalling.
Sigma-Aldrich
Anti-HA (Hemagglutinin) IgG, CF 488A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder