Merck
  • Complete Genome Sequence of Arthrobacter sp. Strain LS16, Isolated from Agricultural Soils with Potential for Applications in Bioremediation and Bioproducts.
Complete Genome Sequence of Arthrobacter sp. Strain LS16, Isolated from Agricultural Soils with Potential for Applications in Bioremediation and Bioproducts.
Genome announcements (2016-01-16)
Ikrema Hassan, Alexander W Eastman, Brian Weselowski, Eltayeb Mohamedelhassan, Ernest K Yanful, Ze-Chun Yuan
ABSTRACT
Here we report the complete genomic sequence of the bacterium Arthrobacter sp. strain LS16, consisting of a single circular chromosome of 3.85 Mb with no identified plasmid. Data contained within will facilitate future genetic modification and engineering of the Arthrobacter sp. LS16 metabolic network to enhance traits relevant to bioremediation and bioproducts.
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 350 purifications