Merck
  • Optimizing imaging speed and excitation intensity for single-molecule localization microscopy.
Optimizing imaging speed and excitation intensity for single-molecule localization microscopy.
Nature methods (2020-08-19)
Robin Diekmann, Maurice Kahnwald, Andreas Schoenit, Joran Deschamps, Ulf Matti, Jonas Ries
ABSTRACT
High laser powers are common practice in single-molecule localization microscopy to speed up data acquisition. Here we systematically quantified how excitation intensity influences localization precision and labeling density, the two main factors determining data quality. We found a strong trade-off between imaging speed and quality and present optimized imaging protocols for high-throughput, multicolor and three-dimensional single-molecule localization microscopy with greatly improved resolution and effective labeling efficiency.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
Anti-AHCTF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution