Merck
  • Altered vitamin D3 metabolism in the ovary and periovarian adipose tissue of rats with letrozole-induced PCOS.
Altered vitamin D3 metabolism in the ovary and periovarian adipose tissue of rats with letrozole-induced PCOS.
Histochemistry and cell biology (2020-10-24)
Malgorzata Grzesiak, Gabriela Burzawa, Patrycja Kurowska, Klaudia Blaszczyk, Agata Szlaga, Anna Blasiak, Andrzej Sechman, Agnieszka Rak
ABSTRACT
Vitamin D3 (VD3) plays an important role in the ovary and its deficiency is associated with ovarian pathologies, including polycystic ovary syndrome (PCOS). However, there is no data related to VD3 metabolism in the ovary during PCOS. Herein, we investigated differences in the expression of VD3 receptor (VDR) and key VD3 metabolic enzymes, 1α-hydroxylase (CYP27B1) and 24-hydroxylase (CYP24A1), in the ovary and periovarian adipose tissue (POAT) of control (proestrus and diestrus) and PCOS induced by letrozole rats. Vdr, Cyp27b1 and Cyp24a1 mRNA expression was determined, their protein abundance was examined and immunolocalized. Furthermore, VD3 metabolite concentrations in plasma (25OHD) and tissues (ovary and POAT; 1,25(OH)2D3), and plasma calcium level were determined. 25OHD concentration decreased markedly in letrozole-treated rats in comparison with controls, whereas calcium concentration did not vary among the examined groups. The amount of 1,25(OH)2D3 decreased in both ovary and POAT of PCOS rats. In the ovary, we found decreased Cyp27b1 and increased Vdr mRNA expression in letrozole-treated and diestrus control group. Corresponding protein abundances were down-regulated and up-regulated, respectively but only following letrozole treatment. In POAT, only Cyp27b1 transcript level and CYP27B1 protein abundance were decreased in letrozole-treated rats. VDR was immunolocalized in healthy and cystic follicles, while CYP27B1 and CYP24A1 were found exclusively in healthy ones. Concluding, our results provide the first evidence of disrupted VD3 metabolism in the ovary and POAT of PCOS rats. The reduced 1,25(OH)2D3 concentration in those tissues suggests their contribution to VD3 deficiency observed in PCOS and might implicate in PCOS pathogenesis.
Sigma-Aldrich
Normal Goat IgG, This Normal Goat IgG is validated for use in ELISA Flow Cytometry Immunoblotting Immunofluorescence Immunohistochemistry Immunoprecipitation for the detection of Goat IgG, Non-immune.
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)