Merck
  • Comprehensive proteomic analysis of Schizosaccharomyces pombe by two-dimensional HPLC-tandem mass spectrometry.
Comprehensive proteomic analysis of Schizosaccharomyces pombe by two-dimensional HPLC-tandem mass spectrometry.
Methods (San Diego, Calif.) (2009-03-11)
Laurence M Brill, Khatereh Motamedchaboki, Shuangding Wu, Dieter A Wolf
ABSTRACT
We describe a detailed and widely applicable method for comprehensive proteomic profiling of the fission yeast Schizosaccharomyces pombe by 2-dimensional high performance liquid chromatography-electrospray ionization-tandem mass spectrometry that demonstrates high sensitivity and robust operation. Steps ranging from the preparation of total proteins, digestion of proteins to peptides, and separation of peptides by two-dimensional (1. strong cation exchange and 2. reversed-phase) high performance liquid chromatography followed by tandem mass spectrometry and data processing have been optimized for our instrumentation platform. Using this technology, we identify ca. 3400 proteins per sample and have identified an estimated 4600 proteins in vegetative cells (equal to ca. 90% of the predicted S. pombe proteome) at a false discovery rate of 0.02. Considering the fact that approximately 500 genes are strongly induced during sexual differentiation, and sexual differentiation was not included in our experiments, the proteomic profiling technique affords what should be virtually complete coverage of the vegetative S. pombe proteome. In addition, these methods are widely applicable, having been used for proteomic profiling of several other organisms.
Sigma-Aldrich
Adenine hemisulfate salt, ≥99%
Pricing and availability is not currently available.