Merck
  • Distinct intra-mitochondrial localizations of pro-survival kinases and regulation of their functions by DUSP5 and PHLPP-1.
Distinct intra-mitochondrial localizations of pro-survival kinases and regulation of their functions by DUSP5 and PHLPP-1.
Biochimica et biophysica acta. Molecular basis of disease (2020-06-02)
Wataru Ohwada, Masaya Tanno, Toshiyuki Yano, Sang-Bing Ong, Koki Abe, Tatsuya Sato, Atsushi Kuno, Takayuki Miki, Hirohito Sugawara, Yusuke Igaki, Tetsuji Miura
ABSTRACT
ERK and Akt have been shown to regulate cell sensitivity to death-inducing stress by phosphorylating GSK-3β, a major modulator of the threshold for mitochondrial permeability transition. Here we examined intra-mitochondrial localization of the pro-survival kinases and their regulation by phosphatases. Stepwise trypsin digestion of mitochondria isolated from HEK293 or H9c2 cells was performed, and immunoblotting revealed that GSK-3β and ERK localized dominantly in the outer membrane (OM), while Akt resided at comparable levels in OM, the inner membrane (IM) and the matrix. Treatment with IGF-1 increased the protein level of Akt in the matrix, while ERK and GSK-3β protein levels were increased in OM. Simultaneously, IGF-1 treatment elevated the level of Thr202/Tyr204-phospho-ERK in IM and matrix and levels of Ser473-phospho-Akt and Ser9-phospho-GSK-3β in OM, IM and matrix. Exposing cells to reactive oxygen species (ROS) by using antimycin A increased the levels of DUSP5 and PHLPP-1 mainly in OM and induced dephosphorylation of Akt, ERK and GSK-3β. The mitochondrial localization of DUSP5 was confirmed by experiments with mitochondria purified by Percoll gradient centrifugation and by transfection of cells with GFP-tagged DUSP5. Knockdown of either DUSP5 or PHLPP-1 increased the levels of both Thr202/Tyr204-phospho-ERK and Ser473-phospho-Akt in mitochondria. Cell death induced by antimycin A was suppressed by siRNA-mediated knockdown of DUSP5. The results suggest that Akt and ERK in mitochondria show distinct intra-mitochondrial localization and crosstalk in GSK-3β regulation and that recruitment of DUSP5 as well as PHLPP-1 to mitochondria contributes to ROS-induced termination of the protective signaling.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
MitoTEMPO, ≥98% (HPLC)
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Tris-Tricine-SDS Buffer 10× Concentrate