Merck
  • A Dual-Mechanism Antibiotic Kills Gram-Negative Bacteria and Avoids Drug Resistance.
A Dual-Mechanism Antibiotic Kills Gram-Negative Bacteria and Avoids Drug Resistance.
Cell (2020-06-05)
James K Martin, Joseph P Sheehan, Benjamin P Bratton, Gabriel M Moore, André Mateus, Sophia Hsin-Jung Li, Hahn Kim, Joshua D Rabinowitz, Athanasios Typas, Mikhail M Savitski, Maxwell Z Wilson, Zemer Gitai
ABSTRACT
The rise of antibiotic resistance and declining discovery of new antibiotics has created a global health crisis. Of particular concern, no new antibiotic classes have been approved for treating Gram-negative pathogens in decades. Here, we characterize a compound, SCH-79797, that kills both Gram-negative and Gram-positive bacteria through a unique dual-targeting mechanism of action (MoA) with undetectably low resistance frequencies. To characterize its MoA, we combined quantitative imaging, proteomic, genetic, metabolomic, and cell-based assays. This pipeline demonstrates that SCH-79797 has two independent cellular targets, folate metabolism and bacterial membrane integrity, and outperforms combination treatments in killing methicillin-resistant Staphylococcus aureus (MRSA) persisters. Building on the molecular core of SCH-79797, we developed a derivative, Irresistin-16, with increased potency and showed its efficacy against Neisseria gonorrhoeae in a mouse vaginal infection model. This promising antibiotic lead suggests that combining multiple MoAs onto a single chemical scaffold may be an underappreciated approach to targeting challenging bacterial pathogens.
Sigma-Aldrich
Irresistin-16, ≥98% (HPLC)
Sigma-Aldrich
Dihydrofolate Reductase Assay Kit, 1 kit sufficient for 50-100 tests