Merck
  • Discovery of thermophilic Bacillales using reduced-representation genotyping for identification.
Discovery of thermophilic Bacillales using reduced-representation genotyping for identification.
BMC microbiology (2020-05-15)
Berenice Talamantes-Becerra, Jason Carling, Andrzej Kilian, Arthur Georges
ABSTRACT
This study demonstrates the use of reduced-representation genotyping to provide preliminary identifications for thermophilic bacterial isolates. The approach combines restriction enzyme digestion and PCR with next-generation sequencing to provide thousands of short-read sequences from across the bacterial genomes. Isolates were obtained from compost, hot water systems, and artesian bores of the Great Artesian Basin. Genomic DNA was double-digested with two combinations of restriction enzymes followed by PCR amplification, using a commercial provider of DArTseq™, Diversity Arrays Technology Pty Ltd. (Canberra, Australia). The resulting fragments which formed a reduced-representation of approximately 2.3% of the genome were sequenced. The sequence tags obtained were aligned against all available RefSeq bacterial genome assemblies by BLASTn to identify the nearest reference genome. Based on the preliminary identifications, a total of 99 bacterial isolates were identified to species level, from which 8 isolates were selected for whole-genome sequencing to assess the identification results. Novel species and strains were discovered within this set of isolates. The preliminary identifications obtained by reduced-representation genotyping, as well as identifications obtained by BLASTn alignment of the 16S rRNA gene sequence, were compared with those derived from the whole-genome sequence data, using the same RefSeq sequence database for the three methods. Identifications obtained with reduced-representation sequencing agreed with the identifications provided by whole-genome sequencing in 100% of cases. The identifications produced by BLASTn alignment of 16S rRNA gene sequence to the same database differed from those provided by whole-genome sequencing in 37.5% of cases, and produced ambiguous identifications in 50% of cases. Previously, this method has been successfully demonstrated for use in bacterial identification for medical microbiology. This study demonstrates the first successful use of DArTseq™ for preliminary identification of thermophilic bacterial isolates, providing results in complete agreement with those obtained from whole-genome sequencing of the same isolates. The growing database of bacterial genome sequences provides an excellent resource for alignment of reduced-representation sequence data for identification purposes, and as the available sequenced genomes continue to grow, the technique will become more effective.
Product Description
Genomic DNA of Escherichia coli O157 (EDL 933), IRMM®, certified reference material
Sigma-Aldrich
Gelzan CM, Gelrite®