Merck
  • ECM-incorporated hydrogels cross-linked via native chemical ligation to engineer stem cell microenvironments.
ECM-incorporated hydrogels cross-linked via native chemical ligation to engineer stem cell microenvironments.
Biomacromolecules (2013-07-24)
Jangwook P Jung, Anthony J Sprangers, John R Byce, Jing Su, Jayne M Squirrell, Phillip B Messersmith, Kevin W Eliceiri, Brenda M Ogle
ABSTRACT
Limiting the precise study of the biochemical impact of whole molecule extracellular matrix (ECM) proteins on stem cell differentiation is the lack of 3D in vitro models that can accommodate many different types of ECM. Here we sought to generate such a system while maintaining consistent mechanical properties and supporting stem cell survival. To this end, we used native chemical ligation to cross-link poly(ethylene glycol) macromonomers under mild conditions while entrapping ECM proteins (termed ECM composites) and stem cells. Sufficiently low concentrations of ECM were used to maintain constant storage moduli and pore size. Viability of stem cells in composites was maintained over multiple weeks. ECM of composites encompassed stem cells and directed the formation of distinct structures dependent on ECM type. Thus, we introduce a powerful approach to study the biochemical impact of multiple ECM proteins (either alone or in combination) on stem cell behavior.
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Laminin-1 A&B chains Antibody, cross region, clone AL-2, clone AL-2, Chemicon®, from rat
Sigma-Aldrich
Anti-Rat Collagen Type I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Actin Cytoskeleton / Focal Adhesion Staining Kit, The Actin Cytoskeleton & Focal Adhesion Staining Kit consists of TRITC-conjugated phalloidin, anti-Vinculin & DAPI for the immunofluorescent staining of actin filaments in the cytoskeleton as well as the nucleus of the cells.