Merck
  • Germinality does not necessarily define mAb expression and thermal stability.
Germinality does not necessarily define mAb expression and thermal stability.
Applied microbiology and biotechnology (2019-07-28)
Linda Schwaigerlehner, Patrick Mayrhofer, Matthias Diem, Willibald Steinfellner, Emma Fenech, Chris Oostenbrink, Renate Kunert
ABSTRACT
The production potential of recombinant monoclonal antibody (mAb) expressing cell lines depends, among other factors, on the intrinsic antibody structure determined by the amino acid sequence. In this study, we investigated the influence of somatic mutations in the V(D)J sequence of four individual, mature model mAbs on the expression potential. Therefore, we defined four couples, each consisting of one naturally occurring mAb (2G12, Ustekinumab, 4B3, and 2F5) and the corresponding germline-derived cognate mAb (353/11, 554/12, 136/63, and 236/14). For all eight mAb variants, recombinant Chinese hamster ovary (CHO) cell lines were developed with mAbs expressed from a defined chromosomal locus. The presented workflow investigates critical parameters including productivity, intra- and extracellular product profile, XBP1 splicing, thermal stability, and in silico hydrophobicity. Significant differences in productivity were even observed between the germline-derived mAbs which did not undergo somatic mutagenesis. Accordingly, back-to-germline mutations of mature mAbs are not necessarily reflecting improved expression and stability but indicate opportunities and limits of mAb engineering. From our studies, we conclude that germinalization represents a potential to improve mAb properties depending on the antibody's germline family, highlighting the fact that mAbs should be treated individually.
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human Lambda Light Chains (Bound and Free)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution