Merck
  • Identification of CRKII, CFL1, CNTN1, NME2, and TKT as Novel and Frequent T-Cell Targets in Human IDH-Mutant Glioma.
Identification of CRKII, CFL1, CNTN1, NME2, and TKT as Novel and Frequent T-Cell Targets in Human IDH-Mutant Glioma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-03-23)
Steffen Dettling, Slava Stamova, Rolf Warta, Martina Schnölzer, Carmen Rapp, Anchana Rathinasamy, David Reuss, Kolja Pocha, Saskia Roesch, Christine Jungk, Uwe Warnken, Volker Eckstein, Niels Grabe, Christoph Schramm, Markus A Weigand, Andreas von Deimling, Andreas Unterberg, Philipp Beckhove, Christel Herold-Mende
ABSTRACT
Purpose: Successful immunotherapies for IDHmut gliomas require better knowledge of T-cell target antigens. Here, we elucidated their antigen repertoire recognized by spontaneous T-cell responses using an unbiased proteomic approach.Experimental Design: Protein fractionations of tissue lysates from IDHmut gliomas (n = 4) were performed. Fractions were tested by IFNγ ELISpot assay for recognition through patients' T cells. Proteins of immunogenic fractions were identified by mass spectrometry and validated by in silico-predicted synthetic long peptides in patients of origin, additional IDHmut glioma patients (n = 16), and healthy donors (n = 13). mRNA and protein expression of immunogenic antigens was analyzed in tumor tissues and IDHmut glioma stem-like cells (GSC). HLA-A*02-restricted T-cell epitopes were functionally determined by short peptides and numbers of antigen-specific T cells by HLA-peptide tetramer analysis.Results: A total of 2,897 proteins were identified in immunogenic tumor fractions. Based on a thorough filter process, 79 proteins were selected as potential T-cell antigens. Twenty-six of these were recognized by the patients' T cells, and five of them (CRKII, CFL1, CNTN1, NME2, and TKT) in up to 56% unrelated IDHmut glioma patients. Most immunogenic tumor-associated antigens (TAA) were expressed in IDHmut gliomas and GSCs, while being almost absent in normal brain tissues. Finally, we identified HLA-A*02-restricted epitopes for CRKII, NME2, and TKT that were recognized by up to 2.82% of antigen-specific peripheral cytotoxic T cells in IDHmut glioma patients.Conclusions: By analyzing the repertoire of T-cell target antigens in IDHmut glioma patients, we identified five novel immunogenic TAAs and confirmed their expression on IDHmut tumors and GSCs. Clin Cancer Res; 24(12); 2951-62. ©2018 AACR.
Sigma-Aldrich
Anti-TKT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial