Merck
  • Context-dependent IL-6 potentiation of interferon- gamma-induced IL-12 secretion and CD40 expression in murine microglia.
Context-dependent IL-6 potentiation of interferon- gamma-induced IL-12 secretion and CD40 expression in murine microglia.
Journal of neurochemistry (2009-08-29)
Hsiao-Wen Lin, Steven W Levison
ABSTRACT
Interleukin-6 (IL-6) is produced by neurons, astrocytes, and microglia, and elevated levels of IL-6 within the CNS have been documented in multiple neurological disorders including Alzheimer's disease, stroke, epilepsy, attention deficit disorder, cerebral palsy, and multiple sclerosis. Here, we sought to understand how IL-6 regulates microglial signal transduction and their immune properties. Using highly enriched cultures of neonatal murine microglia we show that IL-6 alone has direct effects on microglia as it activates STAT3 and extracellular signal-regulated kinase pathways in a time- and dose-dependent fashion and it enhances interferon-gamma (IFNgamma)-stimulated IL-12 secretion. However, other immune properties were only weakly modulated by IL-6 when administered without the soluble IL-6 receptor (sIL-6R). For instance, IFNgamma-induced expression of the co-stimulatory molecule, CD40 was dependent on sIL-6R administration. IL-6 with or without sIL-6R did not affect major histocompatability complex class II expression. In granulocyte-macrophage colony-stimulating factor (GMCSF)-induced dendritic cell-like microglia, IL-6/sIL-6R and IFNgamma stimulated an even greater increase in CD40 expression compared with primary microglia. Altogether, our results demonstrate that microglial responses to IL-6 are not simple in that the effects of IL-6 are context-dependent. In particular, the presence or absence of sIL-6R, IFNgamma or GMCSF will alter the type and amplitude of their response.
Sigma-Aldrich
Random Nonamers, for use as primers in cDNA synthesis, 50 μM in H2O