Tri-O-benzyl-D-glucal
Empirical Formula (Hill Notation):
C27H28O4
CAS Number:
Molecular Weight:
416.51
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1