Skip to Content
Merck
Sodium cyclopentadienylide 2.4 M in THF

Sodium cyclopentadienylide

Sodium cyclopentadienylide 2.4 M in THF
Synonym(s):
Cyclopentadienylsodium solution
Empirical Formula (Hill Notation):
C5H5Na
CAS Number:
Molecular Weight:
88.08

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing