
Simvastatin for system suitability CRS

Synonym(s):
Simvastatin
Empirical Formula (Hill Notation):
C25H38O5
CAS Number:
Molecular Weight:
418.57
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing