
Resorufin β-D-glucuronide sodium salt

Synonym(s):
3-Phenoxazone 7-(β-D-glucuronide)
Linear Formula:
C18H14NO9Na
CAS Number:
Molecular Weight:
411.29
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing