
Potassium hexahydroxoantimonate(V)

Synonym(s):
Potassium antimonate, hydrated, Potassium hexahydroxoantimonate(V)
Linear Formula:
K[Sb(OH)6]
CAS Number:
Molecular Weight:
262.90
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing