Neoxanthin
Synonym(s):
(3S,3′S,5R,5′R,6R,6′S,9′-cis)-6,7-Didehydro-5′,6′-epoxy-5′,6′-dihydro-β,β-carotene-3,3′,5(6H)-triol, 9′-cis-Neoxanthin
Empirical Formula (Hill Notation):
C40H56O4
CAS Number:
Molecular Weight:
600.87
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1