
Mouse ExtrAvidin® Peroxidase Staining Kit antibody produced in goat

Synonym(s):
ExtrAvidin® Peroxidase Staining Kits
Products
Product Category
Brand
Research Area
Post Translational Modification
Showing 1-1 of 1
Compare
Product Number
Description
Pricing