Skip to Content
FJ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
modified Tryptic Soy Agar- Dehydrated Culture Media
Products
Product Category
media (1)
Brand
Millipore (1)
Physical Form
Agency
Quality Segment
Application
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.04323
modified Tryptic Soy Agar- Dehydrated Culture Media,
NutriSelect
®
Prime, with Lecithin and Polysorbate 80, USP, EP, JP, EN 17141, ISO 21149, powder, Suitable for use in Halal and Kosher certified processes
Expand
View Pricing