Milliflex® Pre-filled Media Cassettes
Products
Product Category
Brand
Color
Physical Form
Chemical Composition
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterilization Method
Showing 1-6 of 6
Compare
Product Number
Description
Pricing