
Matrine

Synonym(s):
Matridin-15-one, Sophocarpidine
Empirical Formula (Hill Notation):
C15H24N2O
CAS Number:
Molecular Weight:
248.36
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing