
Maleic acid disodium salt hydrate

Synonym(s):
Sodium maleate hydrate, Sodium toxilate hydrate
Empirical Formula (Hill Notation):
C4H2Na2O4 · xH2O
CAS Number:
Molecular Weight:
160.04 (anhydrous basis)
Products
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing