Skip to Content
Merck
L-3-Aminoisobutyric acid ≥97.0% (TLC)

L-3-Aminoisobutyric acid

L-3-Aminoisobutyric acid ≥97.0% (TLC)
Synonym(s):
(+)-β-Aminoisobutyric acid, (2S)-3-Amino-2-methylpropanoic acid, (S)-β-AIB, (S)-β2-Homoalanine, (S)-3-Amino-2-methylpropionic acid, L-α-Methyl-β-alanine, S-BAIBA
Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12

Products

Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing