
Isosorbide dinitrate

Synonym(s):
ISDN
Empirical Formula (Hill Notation):
C6H8N2O8
CAS Number:
Molecular Weight:
236.14
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing