Skip to Content
Merck
Isocarbophos PESTANAL®, analytical standard

Isocarbophos

Isocarbophos PESTANAL®, analytical standard
Synonym(s):
O-Methyl O-[2-(isopropoxycarbonyl)phenyl] phosphoramidothioate
Empirical Formula (Hill Notation):
C11H16NO4PS
CAS Number:
Molecular Weight:
289.29
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing