Skip to Content
Merck
Fmoc chloride derivatization grade (HPLC), LiChropur™, ≥99.0% (HPLC)

Fmoc chloride

Fmoc chloride derivatization grade (HPLC), LiChropur™, ≥99.0% (HPLC)
Synonym(s):
9-Fluorenylmethoxycarbonyl chloride, 9-Fluorenylmethyl chloroformate, Fmoc-Cl
Empirical Formula (Hill Notation):
C15H11ClO2
CAS Number:
Molecular Weight:
258.70
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Technique
Showing 1-3 of 3