
Dytek® EP diamine

Synonym(s):
1,3-Diaminopentane, 1,3-Pentanediamine
Linear Formula:
CH3CH2CH(NH2)CH2CH2NH2
CAS Number:
Molecular Weight:
102.18
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing