
D-Mannose 6-phosphate disodium salt hydrate

Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Tissue
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing