Skip to Content
Merck
Diisopropyl azodicarboxylate 98%

Diisopropyl azodicarboxylate

Diisopropyl azodicarboxylate 98%
Synonym(s):
DIAD, Diisopropyl azodiformate, Diisopropylazodicarboxylate
Linear Formula:
(CH3)2CHO2CN=NCO2CH(CH3)2
CAS Number:
Molecular Weight:
202.21

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing