Skip to Content
Merck
(DHQD)2Pyr 97%

(DHQD)2Pyr

(DHQD)2Pyr 97%
Synonym(s):
Hydroquinidine-2,5-diphenyl-4,6-pyrimidinediyl diether
Empirical Formula (Hill Notation):
C56H60N6O4
CAS Number:
Molecular Weight:
881.11

Products

Brand
Formula Weight
Melting Point (°C)
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing