
DAF-FM DA

Synonym(s):
4-Amino-5-(N-methylamino)-3′,6′-bis(acetyloxy)-2′,7′-difluoro-spiro[isobenzofuran-1(3H),9′-[9H]xanthen]-3-one, Diaminofluorescein-FM diacetate
Empirical Formula (Hill Notation):
C25H18F2N2O7
CAS Number:
Molecular Weight:
496.42
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing