Skip to Content
FJ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Codeine for system suitability
Synonym(s):
Codeine
Empirical Formula (Hill Notation):
C
18
H
21
NO
3
CAS Number:
76-57-3
Molecular Weight:
299.36
EC Number:
200-969-1
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
Y0001984
Codeine for system suitability,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing