
Citalopram Related Compound B

Synonym(s):
1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-3-hydroxy-1,3-dihydroisobenzofuran-5-carbonitrile oxalate
Empirical Formula (Hill Notation):
C22H23FN2O6
CAS Number:
Molecular Weight:
430.43
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2