
Cisplatin impurity A

Synonym(s):
trans-Platinum(II)diammine dichloride, trans-Diamminedichloroplatinum(II), trans-Dichlorodiammineplatinum(II)
Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing