
Ciprofloxacin impurity standard

Synonym(s):
Ciprofloxacin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C17H18FN3O3 · HCl · H2O
CAS Number:
Molecular Weight:
385.82
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing