
Cholecystokinin Fragment 30-33 Amide

Synonym(s):
CCK-4, Gastrin Tetrapeptide, Tetragastrin, Trp-Met-Asp-Phe amide
Empirical Formula (Hill Notation):
C29H36N6O6S
CAS Number:
Molecular Weight:
596.70
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing