Chiralyst Rh660
Synonym(s):
1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, (R,R)-Et-DUPHOS-Rh, [(R,R)Ethyl-DuPhosRh(COD)]BF4
Empirical Formula (Hill Notation):
C30H48BF4P2Rh
CAS Number:
Molecular Weight:
660.36
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1