
Cetyl alcohol

Synonym(s):
Cetyl alcohol, 1-Hexadecanol, Palmityl alcohol
Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing