Skip to Content
Merck
Cefaclor British Pharmacopoeia (BP) Reference Standard

Cefaclor

Cefaclor British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C15H14ClN3O4S
CAS Number:
Molecular Weight:
367.81
EC Number:

Products

Product Category
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing