
BM Blue POD Substrate, soluble

Synonym(s):
3,3′,5,5′-Tetramethylbenzidine solution
Empirical Formula (Hill Notation):
C16H20N2
CAS Number:
Molecular Weight:
240.34
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing