
Bisphenol A-d16

Synonym(s):
2,2-Bis(4-hydroxyphenyl)propane-d16
Linear Formula:
(CD3)2C(C6D4OD)2
CAS Number:
Molecular Weight:
244.38
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing