
Arabinofuranose

Synonym(s):
L-Arabinofuranose
Empirical Formula (Hill Notation):
C5H10O5
Molecular Weight:
150.13
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing