
Ammonium heptamolybdate tetrahydrate

Synonym(s):
Ammonium heptamolybdate tetrahydrate
Linear Formula:
(NH4)6Mo7O24 · 4 H2O
CAS Number:
Molecular Weight:
1235.86
Products
Product Category
Brand
Formula Weight
Physical Form
Agency
Analyte Suitability
Quality Segment
Technique
Showing 1-3 of 3