
Acetone-2,4-DNPH

Synonym(s):
Acetone-2,4-dinitrophenylhydrazone
Linear Formula:
(CH3)2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing